The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199251.1 | NP_954859.1 | 1 - 2 | 935 | 163 |
GenBank mRNA | AB052939.1 | - | 1 - 2 | 935 | 163 |
BC138133.1 | - | 1 - 2 | 727 | 163 | |
BC138134.1 | - | 1 - 2 | 727 | 163 |