Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tdpoz3
Entrez Gene ID
399674
Gene Name
TD and POZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 93826020 - 93827117 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207271.2 NP_997154.2 1 - 1 351 84

View Full Product Details