Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt72
Entrez Gene ID
105866
Gene Name
keratin 72
Gene Aliases
AI507495, K6irs2, Krt2-35, Krt72-ps
Chromosome Location
Chr.15: 101776560 - 101786458 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213728.1 NP_998893.1 4 - 5 828 115

View Full Product Details