Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dab1
Entrez Gene ID
13131
Gene Name
disabled 1
Gene Aliases
AI956902, C630028C02Rik, scm, scr, scrambler, yot
Chromosome Location
Chr.4: 103619546 - 104744844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177259.4 NP_796233.2 15 - 15 5184 150
NR_104385.1 16 - 16 5454 150
GenBank mRNA AK083206.1 - 2 - 2 3248 150
BE945099.1 - 1 - 1 150

View Full Product Details