The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201643.2 | NP_964005.2 | 1 - 1 | 370 | 138 |
GenBank mRNA | AB094481.1 | - | 1 - 1 | 349 | 138 |
AY227196.1 | - | 1 - 1 | 370 | 138 | |
BC139093.1 | - | 1 - 1 | 350 | 138 | |
BC139094.1 | - | 1 - 1 | 350 | 138 |