Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Haus7
Entrez Gene ID
73738
Gene Name
HAUS augmin-like complex, subunit 7
Gene Aliases
1110020L19Rik, Uchl5ip, Uip1
Chromosome Location
Chr.X: 73437315 - 73459029 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028633.3 NP_082909.1 3 - 4 380 76
NM_207104.1 NP_996987.1 3 - 4 380 76
GenBank mRNA AK050729.1 - 3 - 4 314 76
AY029337.1 - 3 - 4 380 76
BC025464.1 - 3 - 4 371 76

View Full Product Details