Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Raet1d
Entrez Gene ID
56554
Gene Name
retinoic acid early transcript delta
Gene Aliases
RAE-1delta
Chromosome Location
Chr.10: 22361865 - 22374143 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020030.2 NP_064414.1 - 377 144
GenBank mRNA AF257520.1 - 3 - 3 379 144
BC106190.1 - - 377 144

View Full Product Details