Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sfn
Entrez Gene ID
55948
Gene Name
stratifin
Gene Aliases
Er, Mme1, Ywhas
Chromosome Location
Chr.4: 133600556 - 133602168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF058798.1 - 1 - 1 934 84

View Full Product Details