Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pramel4
Entrez Gene ID
347710
Gene Name
preferentially expressed antigen in melanoma like 4
Gene Aliases
C79430, D4Ertd792e
Chromosome Location
Chr.4: 144059126 - 144069324 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF490340.1 - 1 - 1 47 66

View Full Product Details