The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166625.1 | NP_001160097.1 | 4 - 4 | 2230 | 67 |
NM_009913.6 | NP_034043.1 | 3 - 3 | 2162 | 67 | |
GenBank mRNA | AJ131357.2 | - | 4 - 4 | 2167 | 67 |
AK019478.1 | - | - | 2336 | 67 | |
AK050615.1 | - | 3 - 3 | 2163 | 67 | |
AK153975.1 | - | 3 - 3 | 2163 | 67 | |
AK162762.1 | - | 1 - 1 | 106 | 67 |