Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zscan4c
- Entrez Gene ID
-
245109
- Gene Name
- zinc finger and SCAN domain containing 4C
- Gene Aliases
-
Gm397, XM_142517, Zscan4d
- Chromosome Location
-
Chr.7: 11005745 - 11010547 on Build GRCm38
- Gene Symbol
- Zscan4b
- Entrez Gene ID
-
665780
- Gene Name
- zinc finger and SCAN domain containing 4B
- Gene Aliases
-
EG665780
- Chromosome Location
-
Chr.7: 10900740 - 10905050 on Build GRCm38
- Gene Symbol
- Zscan4f
- Entrez Gene ID
-
665902
- Gene Name
- zinc finger and SCAN domain containing 4F
- Gene Aliases
-
C330039H09Rik
- Chromosome Location
-
Chr.7: 11397915 - 11402318 on Build GRCm38
View Full Product Details