Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vwde
Entrez Gene ID
232585
Gene Name
von Willebrand factor D and EGF domains
Gene Aliases
Gm467
Chromosome Location
Chr.6: 13185610 - 13224965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013757.2 NP_001013779.2 3 - 4 587 70
GenBank mRNA BC076621.1 - 3 - 4 587 70
BC151200.1 - 3 - 4 496 70

View Full Product Details