The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017955.2 | NP_001017955.2 | 5 - 6 | 1968 | 79 |
GenBank mRNA | BC064311.1 | - | 3 - 4 | 201 | 79 |
BC094341.1 | - | 1 - 2 | 309 | 79 |