Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uap1l1
Entrez Gene ID
227620
Gene Name
UDP-N-acteylglucosamine pyrophosphorylase 1-like 1
Gene Aliases
5730445F03Rik
Chromosome Location
Chr.2: 25361489 - 25365686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033293.2 NP_001028465.1 6 - 7 1189 82
GenBank mRNA AK077550.1 - 6 - 7 1213 82
AK077912.1 - - 1188 82
AK149178.1 - 6 - 7 1191 82
AK151942.1 - 6 - 7 1190 82
AK159787.1 - 6 - 7 1200 82
AK168977.1 - 6 - 7 1193 82
AK177165.1 - 1 - 2 61 82
AK189248.1 - 1 - 2 61 82
AK195917.1 - 1 - 2 61 82
AK197433.1 - 1 - 2 61 82
AK205071.1 - 1 - 2 61 82
BC043307.1 - - 1185 82
BC068207.1 - 4 - 5 589 82
BC130030.1 - 7 - 8 1172 82

View Full Product Details