Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpcr1
Entrez Gene ID
268949
Gene Name
diffuse panbronchiolitis critical region 1 (human)
Gene Aliases
Gm630
Chromosome Location
Chr.17: 35635755 - 35643695 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033366.3 NP_001028538.1 1 - 2 108 77
GenBank mRNA AK131887.1 - 1 - 2 110 77
AK164929.1 - 1 - 2 109 77
BC150798.1 - 1 - 2 101 77

View Full Product Details