Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbm44
Entrez Gene ID
329207
Gene Name
RNA binding motif protein 44
Gene Aliases
Gm817
Chromosome Location
Chr.1: 91144973 - 91170799 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033408.4 NP_001028580.1 7 - 8 2087 82
GenBank mRNA AK133355.1 - 7 - 8 2088 82
BC150950.1 - 7 - 8 2049 82

View Full Product Details