Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc47a2
Entrez Gene ID
380701
Gene Name
solute carrier family 47, member 2
Gene Aliases
4933429E10Rik
Chromosome Location
Chr.11: 61301627 - 61342860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033542.2 NP_001028714.1 14 - 15 1419 71
GenBank mRNA AK133432.1 - 14 - 15 1420 71
BC147619.1 - 14 - 15 1367 71
BC157913.1 - 14 - 15 1367 71

View Full Product Details