The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033795.4 | NP_001028967.2 | 7 - 7 | 3374 | 110 |
GenBank mRNA | AK136928.1 | - | 1 - 1 | 3064 | 110 |