The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034060.3 | NP_001029232.2 | 8 - 9 | 1363 | 61 |
NM_001170641.1 | NP_001164112.1 | 8 - 9 | 1386 | 61 | |
GenBank mRNA | AK158260.1 | - | 8 - 9 | 1368 | 61 |
BC150907.1 | - | 8 - 9 | 1354 | 61 | |
BC150910.1 | - | 8 - 9 | 1354 | 61 |