The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160239.2 | NP_001153711.1 | 4 - 5 | 399 | 67 |
NM_001190436.1 | NP_001177365.1 | 3 - 4 | 564 | 67 | |
NM_007990.3 | NP_032016.1 | 4 - 5 | 402 | 67 | |
GenBank mRNA | AK002355.1 | - | 4 - 5 | 340 | 67 |
AK008466.1 | - | 3 - 4 | 309 | 67 | |
AK132752.1 | - | 4 - 5 | 345 | 67 | |
AK146359.1 | - | 4 - 5 | 338 | 67 | |
AK151211.1 | - | 4 - 5 | 340 | 67 | |
AK151465.1 | - | 4 - 5 | 340 | 67 | |
AK152738.1 | - | 4 - 5 | 340 | 67 | |
AK168348.1 | - | 4 - 5 | 340 | 67 | |
AV474170.1 | - | 4 - 5 | 402 | 67 | |
AV502224.1 | - | 2 - 3 | 165 | 67 | |
BC058691.1 | - | 4 - 5 | 319 | 67 | |
BC062873.1 | - | 4 - 5 | 331 | 67 | |
BC081463.1 | - | 4 - 5 | 319 | 67 | |
D26610.1 | - | 4 - 5 | 339 | 67 | |
X65922.1 | - | 4 - 5 | 327 | 67 |