Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hs6st2
Entrez Gene ID
50786
Gene Name
heparan sulfate 6-O-sulfotransferase 2
Gene Aliases
6OST2
Chromosome Location
Chr.X: 51386637 - 51681760 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290467.1 NP_001277396.1 2 - 3 946 81
NM_015819.4 NP_056634.3 1 - 2 587 81
GenBank mRNA AK171680.1 - 1 - 2 590 81
BC037659.1 - 1 - 2 538 81
BC047151.1 - 2 - 3 716 81
BC063327.1 - 1 - 2 579 81

View Full Product Details