Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnaset2b
Entrez Gene ID
68195
Gene Name
ribonuclease T2B
Gene Aliases
0610007O07Rik, 4833423A10Rik, 4930532K22Rik, C79750, RNASE6PL, Rnaset2
Chromosome Location
Chr.17: 6978860 - 6998193 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026611.2 NP_080887.1 3 - 4 350 73
GenBank mRNA AK029149.1 - - 304 73
AK084454.1 - - 310 73
BC100330.1 - 3 - 4 275 73

Gene Symbol
Rnaset2a
Entrez Gene ID
100037283
Gene Name
ribonuclease T2A
Gene Aliases
0610007O07Rik, 4833423A10Rik, 4930532K22Rik, C79750, RNASE6PL, Rnaset2
Chromosome Location
Chr.17: 8128592 - 8147827 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083938.2 NP_001077407.1 3 - 4 306 73
GenBank mRNA AK018722.1 - 3 - 4 306 73
AK019234.1 - 3 - 4 298 73
BC031496.1 - 3 - 4 286 73
BC089534.1 - 3 - 4 615 73

View Full Product Details