Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tdg
- Entrez Gene ID
-
21665
- Gene Name
- thymine DNA glycosylase
- Gene Aliases
-
E130317C12Rik, JZA-3, Jza1
- Chromosome Location
-
Chr.10: 82629247 - 82650494 on Build GRCm38
- Gene Symbol
- Gm9855
- Entrez Gene ID
-
624784
- Gene Name
- thymine DNA glycosylase pseudogene
- Gene Aliases
-
ENSMUSG00000051559
- Chromosome Location
-
Chr.10: 42052558 - 42055453 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037190.1 |
|
1 - 1 |
721 |
93 |
GenBank mRNA |
AF069519.1 |
- |
1 - 1 |
682 |
93 |
AK089915.1 |
- |
1 - 1 |
725 |
93 |
View Full Product Details