The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011561.2 | NP_035691.2 | 9 - 10 | 1202 | 92 |
NM_172552.3 | NP_766140.2 | 9 - 10 | 1301 | 92 | |
GenBank mRNA | AK053870.1 | - | 9 - 10 | 1229 | 92 |
AK164090.1 | - | 9 - 10 | 1162 | 92 | |
AK164643.1 | - | 9 - 10 | 1269 | 92 | |
BC085470.1 | - | 9 - 10 | 1248 | 92 | |
BC085471.1 | - | 9 - 10 | 1272 | 92 |