The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178385.3 | NP_848472.2 | 1 - 1 | 1780 | 84 |
GenBank mRNA | AK132036.1 | - | - | 1730 | 84 |