The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008273.2 | NP_032299.1 | 1 - 2 | 852 | 125 |
GenBank mRNA | BC138115.1 | - | 1 - 2 | 852 | 125 |