The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011823.4 | NP_035953.3 | 3 - 3 | 1495 | 124 |
GenBank mRNA | AK021044.1 | - | 1 - 1 | 645 | 124 |
AK033675.1 | - | 3 - 3 | 1477 | 124 | |
AK039041.1 | - | 3 - 3 | 1426 | 124 | |
AK039528.1 | - | 3 - 3 | 1474 | 124 | |
AK140507.1 | - | 3 - 3 | 1407 | 124 |