Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrgprx2
Entrez Gene ID
243978
Gene Name
MAS-related GPR, member X2
Gene Aliases
G370024M05Rik, MrgB10, Mrgprb10
Chromosome Location
Chr.7: 48478619 - 48499270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034868.3 NP_001030040.2 2 - 2 209 142
GenBank mRNA AK148121.1 - 2 - 2 140 142
AK148153.1 - 2 - 2 215 142
BC141225.1 - 2 - 2 132 142
CJ131827.1 - 2 - 2 260 142

View Full Product Details