Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim58
Entrez Gene ID
216781
Gene Name
tripartite motif-containing 58
Gene Aliases
Gm16, Gm1783
Chromosome Location
Chr.11: 58640465 - 58652404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039047.1 NP_001034136.1 1 - 2 421 90

View Full Product Details