The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039364.2 | NP_001034453.1 | 2 - 3 | 263 | 95 |
NM_008614.2 | NP_032640.1 | 3 - 4 | 342 | 95 | |
GenBank mRNA | AK013799.1 | - | 3 - 4 | 343 | 95 |
AK028057.1 | - | 3 - 4 | 342 | 95 | |
AK083103.1 | - | 3 - 4 | 343 | 95 | |
BY257495.1 | - | 2 - 3 | 264 | 95 | |
U81317.1 | - | 2 - 3 | 262 | 95 |