The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039059.1 | NP_001034148.1 | 3 - 3 | 1013 | 69 |
NM_001039060.1 | NP_001034149.1 | 4 - 4 | 1064 | 69 | |
NM_001039061.1 | NP_001034150.1 | 2 - 2 | 1108 | 69 | |
NM_153165.2 | NP_694805.1 | 4 - 4 | 1064 | 69 | |
GenBank mRNA | AK078112.1 | - | 2 - 2 | 1049 | 69 |
AK081546.1 | - | 3 - 3 | 1017 | 69 | |
AK133055.1 | - | 3 - 3 | 70 | ||
AK169500.1 | - | 4 - 4 | 1007 | 69 | |
BC036978.1 | - | 4 - 4 | 1001 | 69 | |
BC141165.1 | - | 3 - 3 | 778 | 69 |