The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011089.2 | NP_035219.2 | 10 - 10 | 2836 | 146 |
GenBank mRNA | AK140776.1 | - | 10 - 10 | 2842 | 146 |
AK172046.1 | - | - | 2837 | 146 |