Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Runx2
Entrez Gene ID
12393
Gene Name
runt related transcription factor 2
Gene Aliases
AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a
Chromosome Location
Chr.17: 44495986 - 44814831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145920.2 NP_001139392.1 3 - 4 1678 129
NM_001146038.2 NP_001139510.1 5 - 6 943 129
NM_001271627.1 NP_001258556.1 4 - 5 1107 129
NM_009820.5 NP_033950.2 5 - 6 910 129
NR_073392.1 4 - 5 694 129
GenBank mRNA AF005936.1 - 4 - 5 1061 129
AF010284.1 - 5 - 6 910 129
AF053954.1 - 4 - 5 524 129
D14636.1 - 3 - 4 1676 129
D14637.1 - 3 - 4 1676 129
DQ458792.1 - 4 - 5 695 129

View Full Product Details