Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vezf1
Entrez Gene ID
22344
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
AI848691, AW046909, db1
Chromosome Location
Chr.11: 88068256 - 88084729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016686.4 NP_057895.2 3 - 4 907 102
GenBank mRNA AF104410.3 - 3 - 4 907 102

View Full Product Details