Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfwd3
Entrez Gene ID
234736
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
BC027246
Chromosome Location
Chr.8: 111270944 - 111300260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146218.4 NP_666330.2 11 - 12 2012 87
GenBank mRNA AK137499.1 - 11 - 12 1992 87
AK137698.1 - 11 - 12 1975 87
AK139032.1 - 11 - 12 1994 87
BC023694.1 - 11 - 12 2017 87
BC026603.1 - 2 - 3 242 87
BC027246.1 - 6 - 7 963 87
BC096602.1 - 11 - 12 1975 87

View Full Product Details