The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033194.2 | NP_001028366.1 | 10 - 11 | 1439 | 94 |
GenBank mRNA | AK145040.1 | - | - | 1441 | 94 |
AK160997.1 | - | 10 - 11 | 1427 | 94 | |
AK165830.1 | - | 10 - 11 | 1440 | 94 | |
BC141157.1 | - | 10 - 11 | 1429 | 94 |