The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037634.3 | NP_001032723.1 | 3 - 4 | 392 | 84 |
GenBank mRNA | AK143411.1 | - | 3 - 4 | 393 | 84 |
BC099498.1 | - | 3 - 4 | 352 | 84 |