The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053111.2 | NP_444341.1 | 2 - 2 | 654 | 82 |
GenBank mRNA | AK157836.1 | - | 2 - 2 | 651 | 82 |
AY316150.1 | - | 2 - 2 | 654 | 82 |