Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acvr1c
Entrez Gene ID
269275
Gene Name
activin A receptor, type IC
Gene Aliases
ACVRLK7, ALK7, Alk-7, C230097P10
Chromosome Location
Chr.2: 58267453 - 58357896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033369.3 NP_001028541.1 4 - 5 655 122
NM_001111030.1 NP_001104500.1 5 - 6 1022 122
GenBank mRNA AK049089.1 - 4 - 5 656 122
AK142396.1 - 5 - 6 1026 122
BC145224.1 - 4 - 5 818 122

View Full Product Details