The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029025.3 | NP_083301.1 | 2 - 2 | 1156 | 146 |
GenBank mRNA | AK015245.1 | - | 2 - 2 | 1157 | 146 |
AK045585.1 | - | 2 - 2 | 1096 | 146 | |
BC095954.1 | - | 1 - 1 | 1169 | 146 | |
BC120863.1 | - | 2 - 2 | 916 | 146 | |
BC132045.1 | - | 2 - 2 | 916 | 146 |