Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg4a
Entrez Gene ID
666468
Gene Name
autophagy related 4A, cysteine peptidase
Gene Aliases
AI627006, AV169859, Apg4a, Atg4al, Autl2
Chromosome Location
Chr.X: 140956917 - 140995441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174875.3 NP_777364.3 - 341 111
GenBank mRNA AK041379.1 - - 340 111
AK162404.1 - - 343 111
BC089500.1 - - 342 111
BY287162.1 - - 350 111
CK023195.1 - - 384 111

View Full Product Details