Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hist1h3d
- Entrez Gene ID
-
319149
- Gene Name
- histone cluster 1, H3d
- Gene Aliases
-
H3-b
- Chromosome Location
-
Chr.13: 23575763 - 23576266 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178204.2 |
NP_835511.1 |
1 - 1 |
389 |
102 |
GenBank mRNA |
Z30939.1 |
- |
1 - 1 |
116 |
102 |
- Gene Symbol
- Hist1h3b
- Entrez Gene ID
-
319150
- Gene Name
- histone cluster 1, H3b
- Gene Aliases
-
H3-53
- Chromosome Location
-
Chr.13: 23752380 - 23752840 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178203.2 |
NP_835510.1 |
1 - 1 |
350 |
102 |
- Gene Symbol
- Hist1h3i
- Entrez Gene ID
-
319153
- Gene Name
- histone cluster 1, H3i
- Gene Aliases
-
H3-H
- Chromosome Location
-
Chr.13: 21782915 - 21783397 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC116383.2 |
- |
1 - 1 |
350 |
102 |
View Full Product Details