Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhfpl3
Entrez Gene ID
269629
Gene Name
lipoma HMGIC fusion partner-like 3
Gene Aliases
A930031L14Rik
Chromosome Location
Chr.5: 22745636 - 23275597 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081231.2 NP_001074700.2 2 - 3 669 95
GenBank mRNA AK033906.1 - 2 - 3 643 95

View Full Product Details