The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081294.1 | NP_001074763.1 | 2 - 3 | 741 | 69 |
GenBank mRNA | BC147639.1 | - | 2 - 3 | 689 | 69 |
BC147641.1 | - | 2 - 3 | 689 | 69 | |
BC147701.1 | - | 2 - 3 | 689 | 69 | |
BC147708.1 | - | 2 - 3 | 689 | 69 |