Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo47
Entrez Gene ID
72973
Gene Name
F-box protein 47
Gene Aliases
2900052P03Rik, Gm1562
Chromosome Location
Chr.11: 97851558 - 97884390 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081435.1 NP_001074904.1 5 - 6 762 95
GenBank mRNA BC158049.1 - 5 - 6 697 95

View Full Product Details