The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081652.1 | NP_001075121.1 | 1 - 2 | 576 | 145 |
GenBank mRNA | BC072589.1 | - | 1 - 2 | 576 | 145 |