Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dytn
Entrez Gene ID
241073
Gene Name
dystrotelin
Gene Aliases
Gm215
Chromosome Location
Chr.1: 63622851 - 63686927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081658.1 NP_001075127.1 14 - 15 2125 66
GenBank mRNA DQ443727.1 - 14 - 15 2125 66

View Full Product Details