Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dytn
Entrez Gene ID
241073
Gene Name
dystrotelin
Gene Aliases
Gm215
Chromosome Location
Chr.1: 63622851 - 63686927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081658.1 NP_001075127.1 7 - 8 853 91
GenBank mRNA AK158076.1 - 4 - 5 4369 91
DQ443727.1 - 7 - 8 853 91

View Full Product Details