Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankfn1
Entrez Gene ID
382543
Gene Name
ankyrin-repeat and fibronectin type III domain containing 1
Gene Aliases
Gm1175
Chromosome Location
Chr.11: 89421012 - 89696936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080933.1 NP_001074402.1 2 - 3 345 65

View Full Product Details