Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr12
Entrez Gene ID
57750
Gene Name
WD repeat domain 12
Gene Aliases
4933402C23Rik, Ytm1, Ytm1p
Chromosome Location
Chr.1: 60076868 - 60098500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199060.1 NP_001185989.1 12 - 13 1664 112
NM_001199061.1 NP_001185990.1 12 - 13 1396 112
NM_021312.5 NP_067287.1 11 - 12 1486 112
GenBank mRNA AB041608.1 - 12 - 13 1665 112
AK007500.1 - 11 - 12 1263 112
AK016611.1 - 12 - 13 1340 112
AK031436.1 - 11 - 12 1249 112
AK149085.1 - 11 - 12 1168 112
AK161299.1 - 12 - 13 1409 112
AY059431.1 - 11 - 12 1177 112
AY059432.1 - 4 - 5 482 112
BC004748.1 - 11 - 12 1320 112
BC052386.1 - 11 - 12 1457 112
BC096651.1 - 11 - 12 1231 112

View Full Product Details